N-Boc-D-prolinol

Leverantör: Apollo Scientific
Varning

Synonyms: (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (R)-1-Boc-2-pyrrolidinemethanol, N-t-Boc-D-prolinol, (R)-(+)-1-Boc-2-pyrrolidinemethanol, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, N-tert-Butoxycarbonyl-D-prolinol, Boc-D-Prolinol

OR315048-1G OR315048-25G OR315048-5G
APOSOR315048-1GEA 585 SEK
APOSOR315048-1G APOSOR315048-25G APOSOR315048-5G
N-Boc-D-prolinol
N-Boc-D-prolinol
Formel: C₁₀H₁₉NO₃
MW: 201,3 g/mol
Smältpunkt: 59…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040580
CAS nummer: 83435-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR