3-Methoxyisonicotinic acid

Leverantör: Apollo Scientific
Varning

OR313056-1G OR313056-250MG
APOSOR313056-1GEA 945 SEK
APOSOR313056-1G APOSOR313056-250MG
3-Methoxyisonicotinic acid
3-Methoxyisonicotinic acid
Formel: C₇H₇NO₃
MW: 153,1354 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07368621
CAS nummer: 654663-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR