2-Aminobenzaldehyde

Leverantör: Apollo Scientific
Varning

OR313018-1G OR313018-5G
APOSOR313018-1GEA 260 SEK
APOSOR313018-1G APOSOR313018-5G
2-Aminobenzaldehyde
2-Aminobenzaldehyde
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokpunkt: 80…85 °C/2mmHg
Smältpunkt: 37 °C
Flampunkt: 113 °C
Storage Temperature: Frys
MDL Number: MFCD00007709
CAS nummer: 529-23-7
EINECS: 208-454-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR