2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Acetaldehydeyl-4,6-dichloropyrimidine
OR313012-1G OR313012-5G OR313012-10G
APOSOR313012-1GEA 260 SEK
APOSOR313012-1G APOSOR313012-5G APOSOR313012-10G
2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde, Technical Grade
2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde
Formel: C₆H₄Cl₂N₂O
MW: 191,0200 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09991563
CAS nummer: 16019-33-3