8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane 97%
Leverantör: Apollo Scientific
Synonyms: tert-Butyl 2-oxa-5,8-diazaspiro[3.5]nonane-8-carboxylate
OR312319-250MG OR312319-100MG
APOSOR312319-250MGEA 8550 SEK
APOSOR312319-250MG APOSOR312319-100MG
8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane 97%
8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane
Formel: C₁₁H₂₀N₂O₃
MW: 228,29 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22035557
CAS nummer: 1367777-12-5