3-Boc-3,9-diazaspiro[5.5]undecane
Leverantör: Apollo Scientific
Synonyms: tert-Butyl 3,9-diazaspiro[5.5]undecane-3-carboxylate
OR312249-1G OR312249-5G
APOSOR312249-1GEA 1125 SEK
APOSOR312249-1G APOSOR312249-5G
3-Boc-3,9-diazaspiro[5.5]undecane
3-Boc-3,9-diazaspiro[5.5]undecane
Formel: C14H26N2O2
MW: 254,3700 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05861627
CAS nummer: 173405-78-2