6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Leverantör: Apollo Scientific
Synonyms: 6-Bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine
OR310898-25G OR310898-100G
APOSOR310898-25GEA 1300 SEK
APOSOR310898-25G APOSOR310898-100G
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
6-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formel: C₆H₅BrN₄
MW: 213.0400 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11656619
CAS nummer: 947248-68-2