3-Cyano-4-pyridineboronic acid pinacol ester

Leverantör: Apollo Scientific
Varning

Synonyms: 3-Cyanopyridine-4-boronic acid, pinacol ester

OR310222-1G OR310222-250MG
APOSOR310222-1GEA 6250 SEK
APOSOR310222-1G APOSOR310222-250MG
3-Cyano-4-pyridineboronic acid pinacol ester
3-Cyano-4-pyridineboronic acid pinacol ester
Formel: C₁₂H₁₅BN₂O₂
MW: 230,07 g/mol
Smältpunkt: 97…99 °C
Storage Temperature: Omgivande
MDL Number: MFCD09037481
CAS nummer: 878194-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR