4-Cyano-3,5-diiodopyridine

Leverantör: Apollo Scientific

OR310220-1G OR310220-500MG
APOSOR310220-1GEA 8550 SEK
APOSOR310220-1G APOSOR310220-500MG
4-Cyano-3,5-diiodopyridine
4-Cyano-3,5-diiodopyridine
Formel: C₆H₂I₂N₂
MW: 355,90 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09037450
CAS nummer: 827616-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR