Uracil-5-boronic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 2,4-Dihydroxypyrimidine-5-boronic acid, (2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid

OR310205-1G OR310205-500MG
APOSOR310205-1GEA 7200 SEK
APOSOR310205-1G APOSOR310205-500MG
Uracil-5-boronic acid
Uracil-5-boronic acid
Formel: C₄H₅BN₂O₄
MW: 155,90 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01318983
CAS nummer: 70523-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR