1-(4,6-Dichloropyridin-3-yl)ethan-1-one
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(4,6-Dichloropyridin-3-yl)ethanone, 5-Acetyl-2,4-dichloropyridine
OR310142-1G OR310142-500MG
APOSOR310142-1GEA 6850 SEK
APOSOR310142-1G APOSOR310142-500MG
1-(4,6-Dichloropyridin-3-yl)ethan-1-one
1-(4,6-Dichloropyridin-3-yl)ethan-1-one
Formel: C₇H₅Cl₂NO
MW: 190,03 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03265322
CAS nummer: 887573-44-6