1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxamide

Leverantör: Apollo Scientific
Varning

OR310118-1G OR310118-250MG
APOSOR310118-1GEA 2900 SEK
APOSOR310118-1G APOSOR310118-250MG
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxamide
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxamide
Formel: C₇H₉N₃O
Smältpunkt: 215…217 °C
Storage Temperature: Omgivande
MDL Number: MFCD11506254
CAS nummer: 851776-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR