2-{1-[(tert-Butoxy)carbonyl]pyrrolidin-2-yl}acetic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

OR310046-1G OR310046-5G
APOSOR310046-1GEA 1100 SEK
APOSOR310046-1G APOSOR310046-5G
2-{1-[(tert-Butoxy)carbonyl]pyrrolidin-2-yl}acetic acid
2-{1-[(tert-Butoxy)carbonyl]pyrrolidin-2-yl}acetic acid
Formel: C₁₁H₁₉NO₄
MW: 229,28 g/mol
Smältpunkt: 112…113 °C
Storage Temperature: Omgivande
MDL Number: MFCD02179026
CAS nummer: 194154-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR