6-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
Leverantör: Apollo Scientific
OR309212-1G OR309212-5G
APOSOR309212-1GEA 3450 SEK
APOSOR309212-1G APOSOR309212-5G
6-Chloroquinoline-2,3-dicarboxylic acid diethyl ester
Formel: C₁₅H₁₄ClNO₄
Storage Temperature: Omgivande
MDL Number: MFCD09787825
CAS nummer: 92525-74-1