(R)-Morpholin-2-ylmethanol hydrochloride
Leverantör: Apollo Scientific
Synonyms: (R)-2-Hydroxymethylmorpholine hydrochloride, (R)-2-Morpholinemethanol hydrochloride
OR308168-1G OR308168-250MG
APOSOR308168-1GEA 1150 SEK
APOSOR308168-1G APOSOR308168-250MG
(R)-Morpholin-2-ylmethanol hydrochloride
(R)-Morpholin-2-ylmethanol hydrochloride
Formel: C₅H₁₁NO₂·HCl
MW: 153,61 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11052541
CAS nummer: 1436436-17-7