(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 95%
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-4-Boc-morpholine-3-carboxylic acid
OR308161-1G OR308161-5G
APOSOR308161-1GEA 1050 SEK
APOSOR308161-1G APOSOR308161-5G
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 95%
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
Formel: C₁₀H₁₇NO₅
MW: 231,25 g/mol
Smältpunkt: 181 °C
Storage Temperature: Frys
MDL Number: MFCD04114909
CAS nummer: 869681-70-9