(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
Leverantör: Apollo Scientific
OR308151-1G OR308151-5G
APOSOR308151-1GEA 855 SEK
APOSOR308151-1G APOSOR308151-5G
(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
Formel: C₁₀H₁₇NO₅
MW: 231,2457 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09260604
CAS nummer: 884512-77-0