(R)-Morpholin-3-ylmethanol hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: (3R)-3-(Hydroxymethyl)morpholine hydrochloride
OR308127-1G OR308127-250MG
APOSOR308127-1GEA 1150 SEK
APOSOR308127-1G APOSOR308127-250MG
(R)-Morpholin-3-ylmethanol hydrochloride
(R)-Morpholin-3-ylmethanol hydrochloride
Formel: C₅H₁₂ClNO₂
MW: 153,6100 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09910295
CAS nummer: 1212377-10-0