Methyl-3-oxocyclobutanecarboxylate
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
Methyl 3-oxocyclobutanecarboxylate
OR308086-100G
OR308086-25G
APOSOR308086-100GEA
1950
SEK
APOSOR308086-100G
APOSOR308086-25G
Methyl-3-oxocyclobutanecarboxylate
Methyl-3-oxocyclobutanecarboxylate
Formel:
C₆H₈O₃ MW: 128,127 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00604222 CAS nummer: 695-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...