6-Chloro-4-quinolinol

Leverantör: Apollo Scientific

Synonyms: 6-Chloroquinolin-4-ol, 6-Chloro-4-hydroxyquinoline

OR307839-1G OR307839-5G
APOSOR307839-1GEA 635 SEK
APOSOR307839-1G APOSOR307839-5G
6-Chloro-4-quinolinol
6-Chloro-4-quinolinol
Formel: C₉H₆ClNO
MW: 179,60 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00024011
CAS nummer: 23432-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR