4-Chloro-2,8-dimethylquinoline
Leverantör: Apollo Scientific
OR307821-1G OR307821-5G
APOSOR307821-1GEA 1475 SEK
APOSOR307821-1G APOSOR307821-5G
4-Chloro-2,8-dimethylquinoline
4-Chloro-2,8-dimethylquinoline
Formel: C₁₁H₁₀ClN
Storage Temperature: Omgivande
MDL Number: MFCD00272352
CAS nummer: 32314-39-9