4-Cyclopropylphenylboronic acid
Leverantör: Apollo Scientific
Varning
OR307250-1G OR307250-250MG
APOSOR307250-1GEA 2375 SEK
APOSOR307250-1G APOSOR307250-250MG
4-Cyclopropylphenylboronic acid
4-Cyclopropylphenylboronic acid
Formel: C₉H₁₁BO₂
Storage Temperature: Omgivande
MDL Number: MFCD01309064
CAS nummer: 302333-80-8