3-Chloro-1,2,4-triazole

Leverantör: Apollo Scientific
Varning

OR307243-10G OR307243-5G
APOSOR307243-10GEA 1725 SEK
APOSOR307243-10G APOSOR307243-5G
3-Chloro-1,2,4-triazole
3-Chloro-1,2,4-triazole
Formel: C₂H₂ClN₃
MW: 103,51 g/mol
Smältpunkt: 169 °C
Storage Temperature: Omgivande
MDL Number: MFCD00128812
CAS nummer: 6818-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR