(1S,2S)-(+)-1,2-Diaminocyclohexane 96%
Leverantör: Apollo Scientific
Fara
Synonyms: (S,S)-DACH, (1S,2S)-Cyclohexane-1,2-diamine
OR307157-100G OR307157-25G OR307157-5G
APOSOR307157-100GEA 3700 SEK
APOSOR307157-100G APOSOR307157-25G APOSOR307157-5G
(1S,2S)-(+)-1,2-Diaminocyclohexane 96%
(1S,2S)-(+)-1,2-Diaminocyclohexane
Chiral intermediate for pharmaceuticals and agrochemicals.
Formel: C₆H₁₄N₂
MW: 114,19 g/mol
Kokpunkt: 104…110 °C/40mmHg
Smältpunkt: 40…45 °C
Densitet: 0,931 g/cm³ (25 °C)
Flampunkt: 70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00062986
CAS nummer: 21436-03-3
EINECS: 606-765-2
UN: 3259
ADR: 8,III