3-(Boc-amino)cyclohexanecarboxylic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid
OR307109-1G OR307109-10G OR307109-5G
APOSOR307109-1GEA 260 SEK
APOSOR307109-1G APOSOR307109-10G APOSOR307109-5G
3-(Boc-amino)cyclohexanecarboxylic acid 97%
3-(Boc-amino)cyclohexanecarboxylic acid
Formel: C₁₂H₂₁NO₄
MW: 243.30 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03453252
CAS nummer: 334932-13-7