N,N-Dimethyl-2-(piperazin-1-yl)benzamideoxalate

Leverantör: Apollo Scientific
Varning

OR30662-1G OR30662-250MG
APOSOR30662-1GEA 3950 SEK
APOSOR30662-1G APOSOR30662-250MG
N,N-Dimethyl-2-(piperazin-1-yl)benzamideoxalate
N,N-Dimethyl-2-(piperazin-1-yl)benzamideoxalate
Formel: C₁₅H₂₁N₃O₅
Storage Temperature: Omgivande
CAS nummer: 494782-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR