N1-Methylbenzene-1,2-diamine dihydrochloride
Leverantör: Apollo Scientific
Fara
Synonyms: N-Methyl-1,2-benzenediamine dihydrochloride, N-Methyl-1,2-phenylenediamine dihydrochloride
OR30623-100G OR30623-25G
APOSOR30623-100GEA 190 SEK
APOSOR30623-100G APOSOR30623-25G
N1-Methylbenzene-1,2-diamine dihydrochloride
N1-Methylbenzene-1,2-diamine dihydrochloride
Formel: C₇H₁₂Cl₂N₂
MW: 195,0900 g/mol
Smältpunkt: 192 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042021
CAS nummer: 25148-68-9
UN: 2811
ADR: 6.1,III