Thieno[3,2-c]pyridine
Leverantör: Apollo Scientific
Varning
OR30575-100MG OR30575-250MG
APOSOR30575-100MGEA 210 SEK
APOSOR30575-100MG APOSOR30575-250MG
Thieno[3,2-c]pyridine
Thieno[3,2-c]pyridine
Formel: C₇H₅NS
MW: 135,1863 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD01760493
CAS nummer: 272-14-0
EINECS: 205-984-7