6-Bromo-1H-pyrazolo[4,3-b]pyridine
Leverantör: Apollo Scientific
Varning
OR305328-10G OR305328-1G OR305328-5G
APOSOR305328-10GEA 3125 SEK
APOSOR305328-10G APOSOR305328-1G APOSOR305328-5G
6-Bromo-1H-pyrazolo[4,3-b]pyridine
6-Bromo-1H-pyrazolo[4,3-b]pyridine
Formel: C₆H₄BrN₃
MW: 198,0201 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD12024534
CAS nummer: 1150617-54-1