Isoindolin-1-one

Leverantör: Apollo Scientific
Varning

Synonyms: 2,3-Dihydro-1H-isoindol-1-one, 1-Isoindolinone

OR305294-1G OR305294-5G
APOSOR305294-1GEA 500 SEK
APOSOR305294-1G APOSOR305294-5G
Isoindolin-1-one
Isoindolin-1-one
Formel: C₈H₇NO
MW: 133,15 g/mol
Kokpunkt: 203…204 °C/18mmHg
Smältpunkt: 148…153 °C
Storage Temperature: Omgivande
MDL Number: MFCD03085939
CAS nummer: 480-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR