4-Isobutylphenylboronic acid pinacol ester

Leverantör: Apollo Scientific

Synonyms: 4-Isobutylphenylboronic acid, pinacol ester, 2-(4-Isobutylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303958-1G OR303958-500MG
APOSOR303958-1GEA 2200 SEK
APOSOR303958-1G APOSOR303958-500MG
4-Isobutylphenylboronic acid pinacol ester
4-Isobutylphenylboronic acid pinacol ester
Formel: C₁₆H₂₅BO₂
MW: 260,1795 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05663842
CAS nummer: 1033753-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR