2,4-Dichloro-3-formylpyridine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2,4-Dichloronicotinaldehyde, 2,4-Dichloropyridine-3-carboxaldehyde, 2,4-Dichloro-3-formylpyridine, 2,4-Dichloro-3-pyridinecarboxaldehyde
OR303843-1G OR303843-5G OR303843-25G
APOSOR303843-1GEA 1050 SEK
APOSOR303843-1G APOSOR303843-5G APOSOR303843-25G
2,4-Dichloro-3-formylpyridine 98%
2,4-Dichloro-3-formylpyridine
Formel: C₆H₃Cl₂NO
MW: 176,00 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07437909
CAS nummer: 134031-24-6