5-Methoxy-4-methylpyridin-3-amine hydrochloride
Leverantör: Apollo Scientific
OR303840-1G OR303840-500MG
APOSOR303840-1GEA 7900 SEK
APOSOR303840-1G APOSOR303840-500MG
5-Methoxy-4-methylpyridin-3-amine hydrochloride
5-Methoxy-4-methylpyridin-3-amine hydrochloride
Formel: C₇H₁₁ClN₂O
Storage Temperature: Omgivande
MDL Number: MFCD11857675
CAS nummer: 1185094-00-1