Ethyl-4-chloro-6-methylpyridine-2-carboxylate
Leverantör: Apollo Scientific
Varning
OR303783-1G OR303783-500MG
APOSOR303783-1GEA 4275 SEK
APOSOR303783-1G APOSOR303783-500MG
Ethyl-4-chloro-6-methylpyridine-2-carboxylate
Ethyl-4-chloro-6-methylpyridine-2-carboxylate
Formel: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09839163
CAS nummer: 315494-03-2