(2,3-Dimethoxyphenyl)(4-pyridinyl)methanol
Leverantör: Apollo Scientific
OR303707-1G OR303707-500MG
APOSOR303707-1GEA 2925 SEK
APOSOR303707-1G APOSOR303707-500MG
(2,3-Dimethoxyphenyl)(4-pyridinyl)methanol
(2,3-Dimethoxyphenyl)(4-pyridinyl)methanol
Formel: C₁₄H₁₅NO₃
Storage Temperature: Omgivande
MDL Number: MFCD04966948
CAS nummer: 243640-27-9