3-Chlorophenylboronic acid pinacol ester

Leverantör: Apollo Scientific

Synonyms: 2-(3-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chlorophenylboronic acid, pinacol ester

OR303551-10G OR303551-1G OR303551-5G
APOSOR303551-10GEA 1375 SEK
APOSOR303551-10G APOSOR303551-1G APOSOR303551-5G
3-Chlorophenylboronic acid pinacol ester
3-Chlorophenylboronic acid pinacol ester
Formel: C₁₂H₁₆BClO₂
MW: 238,52 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05663877
CAS nummer: 635305-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR