9,9-Dimethylfluoren-2-boronic acid 97%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
(9,9-Dimethyl-9H-fluoren-2-yl)boronic acid
OR303506-1G
OR303506-5G
OR303506-25G
APOSOR303506-1GEA
600
SEK
APOSOR303506-1G
APOSOR303506-5G
APOSOR303506-25G
9,9-Dimethylfluoren-2-boronic acid 97%
9,9-Dimethylfluoren-2-boronic acid
Formel:
C₁₅H₁₅BO₂ MW: 238,09 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD08704227 CAS nummer: 333432-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...