4-(9-Carbazolyl)benzeneboronic acid pinacol ester

Leverantör: Apollo Scientific

Synonyms: 9-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole, 9-Carbazolylbenzeneboronic acid pinacol ester

OR303496-1G OR303496-5G
APOSOR303496-1GEA 740 SEK
APOSOR303496-1G APOSOR303496-5G
4-(9-Carbazolyl)benzeneboronic acid pinacol ester
4-(9-Carbazolyl)benzeneboronic acid pinacol ester
Formel: C₂₄H₂₄BNO₂
MW: 369,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16294549
CAS nummer: 785051-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR