2-Chlorobenzene-1-carboximidamide
Leverantör: Apollo Scientific
|
Varning
|
OR303066-1G
OR303066-500MG
APOSOR303066-1GEA
4875
SEK
APOSOR303066-1G
APOSOR303066-500MG
2-Chlorobenzene-1-carboximidamide
2-Chlorobenzene-1-carboximidamide
Formel:
C₇H₇ClN₂ Storage Temperature: Omgivande |
MDL Number:
MFCD05662890 CAS nummer: 45743-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...