4-Amino-3-furazanecarboxamidoxime

Leverantör: Apollo Scientific
Varning

Synonyms: 4-Amino-N-hydroxy-1,2,5-oxadiazole-3-carboxamidine, 4-Amino-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide

OR303000-1G OR303000-500MG
APOSOR303000-1GEA 2675 SEK
APOSOR303000-1G APOSOR303000-500MG
4-Amino-3-furazanecarboxamidoxime
4-Amino-3-furazanecarboxamidoxime
Formel: C₃H₅N₅O₂
MW: 143,11 g/mol
Smältpunkt: 189…193 °C
Storage Temperature: Omgivande
MDL Number: MFCD00186407
CAS nummer: 13490-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR