2-Bromophenyl isocyanate 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 1-Bromo-2-isocyanatobenzene
OR30274-1G OR30274-25G OR30274-5G
APOSOR30274-1GEA 175 SEK
APOSOR30274-1G APOSOR30274-25G APOSOR30274-5G
2-Bromophenyl isocyanate 97%
2-Bromophenyl isocyanate
  • May form small amount of sediment.
Formel: C₇H₄BrNO
MW: 198,02 g/mol
Kokpunkt: 91…92 °C/5mmHg
Densitet: 1,607 g/cm³ (25 °C)
Flampunkt: 108 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001995
CAS nummer: 1592-00-3
UN: 2206
ADR: 6.1,III