2-Methoxyphenyl isothiocyanate 98%
Leverantör: Apollo Scientific
OR30227-25G OR30227-5G
APOSOR30227-25GEA 2150 SEK
APOSOR30227-25G APOSOR30227-5G
2-Methoxyphenyl isothiocyanate 98%
2-Methoxyphenyl isothiocyanate
Formel: C₈H₇NOS
MW: 165.22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011675
CAS nummer: 3288-04-8