1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazole-4-boronic acid Pinacol ester
OR302141-1G OR302141-5G
APOSOR302141-1GEA 515 SEK
APOSOR302141-1G APOSOR302141-5G
1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel: C₁₄H₂₃BN₂O₃
MW: 278.1550 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD12033229
CAS nummer: 1040377-03-4