3,5-Dimethoxyphenyl isothiocyanate

Leverantör: Apollo Scientific
Fara

Synonyms: 1,3-Dimethoxy-5-isothiocyanatobenzene

OR30188-1G OR30188-5G
APOSOR30188-1GEA 1475 SEK
APOSOR30188-1G APOSOR30188-5G
3,5-Dimethoxyphenyl isothiocyanate
3,5-Dimethoxyphenyl isothiocyanate
Formel: C₉H₉NO₂S
Kokpunkt: 130…132 °C/3mmHg
Smältpunkt: 49…52 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041077
CAS nummer: 104968-58-3
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR