2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
Leverantör: Apollo Scientific
Varning
Synonyms: 2,2-Dimethyl-1,3-dioxane-4,6-dione
OR30175-1KG OR30175-250G OR30175-25G
APOSOR30175-1KGEA 1750 SEK
APOSOR30175-1KG APOSOR30175-250G APOSOR30175-25G
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
Formel: C₆H₈O₄
MW: 144,13 g/mol
Smältpunkt: 92…96 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006638
CAS nummer: 2033-24-1
EINECS: 217-992-8