Thiazole-5-boronic acid pinacol ester 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Thiazole-5-boronic Acid Pinacol Ester
OR301381-250MG OR301381-1G OR301381-5G
APOSOR301381-250MGEA 615 SEK
APOSOR301381-250MG APOSOR301381-1G APOSOR301381-5G
Thiazole-5-boronic acid pinacol ester 97%
Thiazole-5-boronic acid pinacol ester
Formel: C₉H₁₄BNO₂S
MW: 211.09 g/mol
Smältpunkt: 98 °C
Storage Temperature: Frys
MDL Number: MFCD10697440
CAS nummer: 1086111-09-2