6-Methyl-2-(4-methylphenyl)imidazol[1,2-a]pyridine-3-acetic acid 98%
Leverantör: Apollo Scientific
OR301232-10G OR301232-5G OR301232-25G
APOSOR301232-10GEA 1625 SEK
APOSOR301232-10G APOSOR301232-5G APOSOR301232-25G
6-Methyl-2-(4-methylphenyl)imidazol[1,2-a]pyridine-3-acetic acid 98%
6-Methyl-2-(4-methylphenyl)imidazol[1,2-a]pyridine-3-acetic acid
Formel: C₁₇H₁₆N₂O₂
MW: 280,3211 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04114705
CAS nummer: 189005-44-5