N-(3-Aminopropyl)methacrylamide hydrochloride 95%
Leverantör: Apollo Scientific
Varning
OR301178-5G OR301178-10G OR301178-25G
APOSOR301178-5GEA 960 SEK
APOSOR301178-5G APOSOR301178-10G APOSOR301178-25G
N-(3-Aminopropyl)methacrylamide hydrochloride 95%
N-(3-Aminopropyl)methacrylamide hydrochloride
Formel: C₇H₁₅ClN₂O
MW: 178.6598 g/mol
Smältpunkt: 123…128 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00078257
CAS nummer: 72607-53-5