1-(tert-Butoxycarbonyl)-3,3-dimethyl-4-oxopiperidine 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,3-Dimethylpiperidin-4-one, N-BOC protected
OR301152-100G OR301152-25G OR301152-5G
APOSOR301152-100GEA 5900 SEK
APOSOR301152-100G APOSOR301152-25G APOSOR301152-5G
1-(tert-Butoxycarbonyl)-3,3-dimethyl-4-oxopiperidine 95%
1-(tert-Butoxycarbonyl)-3,3-dimethyl-4-oxopiperidine
Formel: C₁₂H₂₁NO₃
MW: 227,3000 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09880282
CAS nummer: 324769-06-4