Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid Pinacol ester, Benzyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1-Cbz-1,2,3,6-tetrahydropyridine-4-boronic acid Pinacol ester, 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, Benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1-Carbobenzoxy-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-[1-(Carbobenzoxy)-1,2,3,6-tetrahydro-4-pyridyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-Cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester
OR301116-1G OR301116-5G
APOSOR301116-1GEA 910 SEK
APOSOR301116-1G APOSOR301116-5G
Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 97%
Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate
Formel: C₁₉H₂₆BNO₄
MW: 343,23 g/mol
Smältpunkt: 93…97 °C
Storage Temperature: Kylskåp
MDL Number: MFCD11521562
CAS nummer: 286961-15-7